siRNA Details
| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172987.2 | NP_766575.2 | 5 | 2052 | ||
| GenBank mRNA | AK049776.1 | - | 5 | 2252 | ||
| BC075683.1 | - | 5 | 2052 | 30613986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkain3| Gene Symbol: Nkain3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172987.2 | NP_766575.2 | 5 | 2052 | ||
| GenBank mRNA | AK049776.1 | - | 5 | 2252 | ||
| BC075683.1 | - | 5 | 2052 | 30613986 | ||