siRNA Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172994.2 | NP_766582.1 | 9 | 3231 | ||
| GenBank mRNA | AK032023.1 | - | 9 | 3134 | ||
| AY348866.1 | - | 9 | 3233 | |||
| BC059811.1 | - | 9 | 3231 | 6849535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r2c| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172994.2 | NP_766582.1 | 9 | 3231 | ||
| GenBank mRNA | AK032023.1 | - | 9 | 3134 | ||
| AY348866.1 | - | 9 | 3233 | |||
| BC059811.1 | - | 9 | 3231 | 6849535 | ||