siRNA Details
| Gene Symbol: ZBED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171135.1 | NP_001164606.1 | 2 | 2500 | ||
| NM_001171136.1 | NP_001164607.1 | 2 | 2422 | |||
| NM_004729.3 | NP_004720.1 | 2 | 2419 | |||
| GenBank mRNA | AB018328.1 | - | 2 | 2419 | ||
| BC015030.1 | - | 2 | 2500 | 3908412 | ||
| DA625185.1 | - | 1 | 81 | |||
| Y16947.1 | - | 1 | 2274 | |||

ZBED1
Genomic Map