siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 4 | 490 | ||
| GenBank mRNA | AF212319.1 | - | 4 | 461 | ||
| AK088110.1 | - | 4 | 462 | |||
| AK145753.1 | - | 4 | 435 | |||
| AK146564.1 | - | 4 | 473 | |||
| AK161640.1 | - | 4 | 474 | |||
| AK169395.1 | - | 4 | 487 | |||
| AK181957.1 | - | 2 | 197 | |||
| AK212860.1 | - | 2 | 195 | |||
| BC060030.1 | - | 4 | 478 | 6515657 | ||

Idh2
Genomic Map