siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 6, 7 | 898 | ||
| GenBank mRNA | AF212319.1 | - | 6, 7 | 869 | ||
| AK088110.1 | - | 6, 7 | 870 | |||
| AK145753.1 | - | 6, 7 | 843 | |||
| AK146564.1 | - | 6, 7 | 984 | |||
| AK161640.1 | - | 6, 7 | 882 | |||
| AK169395.1 | - | 6, 7 | 895 | |||
| AK190762.1 | - | 111 | ||||
| BC060030.1 | - | 6, 7 | 886 | 6515657 | ||

Idh2
Genomic Map