siRNA Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048143.2 | NP_001041608.1 | 6 | 2636 | ||
| NM_173015.3 | NP_766603.1 | 5 | 2826 | |||
| GenBank mRNA | AK030288.1 | - | 5 | 2792 | ||
| AK157390.1 | - | 6 | 2642 | |||
| AK173047.1 | - | 5 | 2498 | |||
| BC062639.1 | - | 5 | 2502 | |||

Mon1b
Genomic Map