siRNA Details
| Gene Symbol: Catsperb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173023.2 | NP_766611.2 | 23, 24 | 2953 | ||
| GenBank mRNA | AK029843.1 | - | 23, 24 | 2955 | ||
| AK030010.1 | - | 23, 24 | 2944 | |||
| BC132479.1 | - | 23, 24 | 2874 | 40130756 | ||
| BC145000.1 | - | 22, 23 | 2729 | |||
| EF199807.1 | - | 22, 23 | 2834 | |||

Catsperb
Genomic Map