siRNA Details
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 48 | 6836 | ||
| GenBank mRNA | AB115421.1 | - | 48 | 6831 | ||
| AK029649.1 | - | 2 | 591 | |||
| AK030073.1 | - | 1 | 60 | |||
| AK135983.1 | - | 5 | 990 | |||
| AK220362.1 | - | 1 | 253 | |||
| BC062130.1 | - | 4 | 734 | |||
| BC096768.1 | - | 2 | 430 | |||
| BC145012.1 | - | 48 | 6725 | |||

Vps13a
Genomic Map