siRNA Details
| Gene Symbol: Zfp445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173364.5 | NP_775540.4 | 7 | 2643 | ||
| GenBank mRNA | AK036132.1 | - | 1 | 806 | ||
| AY341877.1 | - | 6 | 2484 | |||
| BC027167.1 | - | 1 | 1389 | |||
| BC034572.1 | - | 1 | 1253 | |||
| BC055684.1 | - | 1 | 940 | |||
| BC099896.1 | - | 6 | 2455 | 30545532 | ||
| BC099930.1 | - | 7 | 2588 | 30058096 | ||

Zfp445
Genomic Map