siRNA Details
Gene Symbol: Dleu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173419.2 | NP_775595.1 | 2 | 737 | ||
GenBank mRNA | AK082171.1 | - | 3 | 499 | ||
BC038059.1 | - | 2 | 730 | 4235022 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dleu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173419.2 | NP_775595.1 | 2 | 737 | ||
GenBank mRNA | AK082171.1 | - | 3 | 499 | ||
BC038059.1 | - | 2 | 730 | 4235022 |