siRNA Details
| Gene Symbol: Klhdc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173427.2 | NP_775603.2 | 1 | 2619 | ||
| GenBank mRNA | AK045732.1 | - | 1 | 2622 | ||
| AK050048.1 | - | 1 | 2206 | |||
| AK052747.1 | - | 1 | 2188 | |||
| BC043462.1 | - | 1 | 1158 | |||
| BC043672.1 | - | 1 | 1158 | |||

Klhdc7a
Genomic Map