siRNA Details
| Gene Symbol: Zfp775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173429.2 | NP_775605.1 | 2 | 723 | ||
| GenBank mRNA | AK048054.1 | - | 2 | 723 | ||
| AK163249.1 | - | 3 | 829 | |||
| AK167452.1 | - | 3 | 829 | |||
| BC141223.1 | - | 2 | 718 | 9055835 | ||

Zfp775
Genomic Map