siRNA Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173446.2 | NP_775622.1 | 1 | 891 | ||
| GenBank mRNA | AK032202.1 | - | 1 | 891 | ||
| AK075681.1 | - | 1 | 21 | |||
| BC117535.1 | - | 1 | 889 | 40106181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155a| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173446.2 | NP_775622.1 | 1 | 891 | ||
| GenBank mRNA | AK032202.1 | - | 1 | 891 | ||
| AK075681.1 | - | 1 | 21 | |||
| BC117535.1 | - | 1 | 889 | 40106181 | ||