siRNA Details
| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173739.3 | NP_776100.2 | 5 | 1251 | ||
| GenBank mRNA | AB175685.1 | - | 5 | 821 | ||
| BC024988.1 | - | 5 | 1251 | 5044626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt18| Gene Symbol: Galnt18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173739.3 | NP_776100.2 | 5 | 1251 | ||
| GenBank mRNA | AB175685.1 | - | 5 | 821 | ||
| BC024988.1 | - | 5 | 1251 | 5044626 | ||