siRNA Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173767.3 | NP_776128.2 | 9 | 1418 | ||
| GenBank mRNA | AK050143.1 | - | 6 | 913 | ||
| AK178410.1 | - | 1 | 92 | |||
| AK196619.1 | - | 1 | 92 | |||
| BC026405.1 | - | 9 | 1305 | 4211657 | ||
| BC089586.1 | - | 6 | 922 | |||
| BC139196.1 | - | 6 | 806 | 8862218 | ||
| BC139197.1 | - | 6 | 806 | 8862219 | ||
| DQ205645.1 | - | 8 | 1131 | |||

Insc
Genomic Map