siRNA Details
| Gene Symbol: Neu4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173772.3 | NP_776133.1 | 4 | 3636 | ||
| GenBank mRNA | AK047397.1 | - | 1 | 1812 | ||
| BC070438.1 | - | 4 | 3636 | 6842155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neu4| Gene Symbol: Neu4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173772.3 | NP_776133.1 | 4 | 3636 | ||
| GenBank mRNA | AK047397.1 | - | 1 | 1812 | ||
| BC070438.1 | - | 4 | 3636 | 6842155 | ||