siRNA Details
| Gene Symbol: Slc45a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173774.3 | NP_776135.2 | 9 | 2520 | ||
| GenBank mRNA | AK082651.1 | - | 2513 | |||
| AK134683.1 | - | 9 | 2511 | |||
| BC051188.1 | - | 9 | 2482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc45a1| Gene Symbol: Slc45a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173774.3 | NP_776135.2 | 9 | 2520 | ||
| GenBank mRNA | AK082651.1 | - | 2513 | |||
| AK134683.1 | - | 9 | 2511 | |||
| BC051188.1 | - | 9 | 2482 | |||