siRNA Details
| Gene Symbol: Sh2d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173778.3 | NP_776139.1 | 3 | 589 | ||
| GenBank mRNA | AK030178.1 | - | 3 | 590 | ||
| BC116842.1 | - | 3 | 512 | 40126161 | ||
| BC116844.1 | - | 3 | 512 | 40126163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2d7| Gene Symbol: Sh2d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173778.3 | NP_776139.1 | 3 | 589 | ||
| GenBank mRNA | AK030178.1 | - | 3 | 590 | ||
| BC116842.1 | - | 3 | 512 | 40126161 | ||
| BC116844.1 | - | 3 | 512 | 40126163 | ||