siRNA Details
| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173863.2 | NP_776288.2 | 15 | 1919 | ||
| GenBank mRNA | AK144840.1 | - | 15 | 1922 | ||
| BC011210.1 | - | 15 | 1758 | 4195538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtc3| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173863.2 | NP_776288.2 | 15 | 1919 | ||
| GenBank mRNA | AK144840.1 | - | 15 | 1922 | ||
| BC011210.1 | - | 15 | 1758 | 4195538 | ||