siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175031.3 | NP_778196.2 | 10 | 1464 | ||
| GenBank mRNA | AK029842.1 | - | 10 | 1381 | ||
| AK173142.1 | - | 9 | 1330 | |||
| BC043103.1 | - | 10 | 1416 | 6406485 | ||
| BC058698.1 | - | 10 | 1529 | 6831702 | ||
| BC061470.1 | - | 9 | 1273 | |||

Stk36
Genomic Map