siRNA Details
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175484.3 | NP_780693.2 | 5 | 994 | ||
GenBank mRNA | AK047527.1 | - | 5 | 936 | ||
AK087414.1 | - | 5 | 995 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175484.3 | NP_780693.2 | 5 | 994 | ||
GenBank mRNA | AK047527.1 | - | 5 | 936 | ||
AK087414.1 | - | 5 | 995 |