siRNA Details
| Gene Symbol: 6430571L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175486.3 | NP_780695.1 | 3 | 541 | ||
| GenBank mRNA | AK032358.1 | - | 3 | 542 | ||
| AK032501.1 | - | 3 | 548 | |||
| AK043338.1 | - | 3 | 581 | |||
| AK085251.1 | - | 3 | 572 | |||
| BC115775.1 | - | 3 | 453 | 40107115 | ||
| BC116708.1 | - | 3 | 453 | 40107114 | ||

6430571L13Rik
Genomic Map