siRNA Details
| Gene Symbol: Pnma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175498.4 | NP_780707.1 | 3 | 3403 | ||
| GenBank mRNA | AK084459.1 | - | 1 | 1592 | ||
| AK164540.1 | - | 1 | 1561 | |||
| AK220357.1 | - | 3362 | ||||
| BC065116.1 | - | 2 | 3234 | 30618233 | ||

Pnma2
Genomic Map