siRNA Details
| Gene Symbol: Adamts12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175501.2 | NP_780710.2 | 2 | 594 | ||
| GenBank mRNA | AJ537452.1 | - | 2 | 353 | ||
| AK048612.1 | - | 2 | 557 | |||
| AK054015.1 | - | 2 | 566 | |||
| AK135354.1 | - | 2 | 596 | |||
| BC151248.1 | - | 2 | 357 | |||
| BC152338.1 | - | 2 | 357 | |||

Adamts12
Genomic Map