siRNA Details
| Gene Symbol: Adamts12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175501.2 | NP_780710.2 | 3 | 786 | ||
| GenBank mRNA | AJ537452.1 | - | 3 | 545 | ||
| AK048612.1 | - | 3 | 749 | |||
| AK135354.1 | - | 3 | 788 | |||
| BC151248.1 | - | 3 | 549 | |||
| BC152338.1 | - | 3 | 549 | |||

Adamts12
Genomic Map