siRNA Details
| Gene Symbol: Lingo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165999.1 | NP_001159471.1 | 8 | 1507 | ||
| NM_001166000.1 | NP_001159472.1 | 9 | 966 | |||
| NM_001166001.1 | NP_001159473.1 | 2 | 427 | |||
| NM_175516.4 | NP_780725.2 | 3 | 534 | |||
| GenBank mRNA | AK035778.1 | - | 3 | 476 | ||
| AK045639.1 | - | 3 | 535 | |||
| AK141568.1 | - | 8 | 990 | |||
| BC090619.1 | - | 9 | 894 | 6734654 | ||
| BC137866.1 | - | 2 | 368 | 8860885 | ||
| BC144985.1 | - | 2 | 368 | 9053533 | ||
| BC145692.1 | - | 2 | 368 | 40130995 | ||

Lingo2
Genomic Map