siRNA Details
| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175526.3 | NP_780735.2 | 6 | 1975 | ||
| GenBank mRNA | AK031121.1 | - | 6 | 1795 | ||
| AK036846.1 | - | 6 | 1780 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec1a| Gene Symbol: Clec1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175526.3 | NP_780735.2 | 6 | 1975 | ||
| GenBank mRNA | AK031121.1 | - | 6 | 1795 | ||
| AK036846.1 | - | 6 | 1780 | |||