siRNA Details
| Gene Symbol: Rttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175542.3 | NP_780751.2 | 20, 21 | 2942 | ||
| GenBank mRNA | AF265232.1 | - | 20, 21 | 2942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rttn| Gene Symbol: Rttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175542.3 | NP_780751.2 | 20, 21 | 2942 | ||
| GenBank mRNA | AF265232.1 | - | 20, 21 | 2942 | ||