siRNA Details
| Gene Symbol: Wdr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175552.4 | NP_780761.1 | 2 | 238 | ||
| GenBank mRNA | AK043557.1 | - | 2 | 226 | ||
| AK050738.1 | - | 2 | 206 | |||
| AK050797.1 | - | 2 | 209 | |||
| BC063100.1 | - | 2 | 224 | 6332749 | ||
| BY125127.1 | - | 2 | 239 | |||

Wdr3
Genomic Map