siRNA Details
| Gene Symbol: Clspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175554.4 | NP_780763.3 | 2, 3 | 259 | ||
| GenBank mRNA | AK045076.1 | - | 2, 3 | 255 | ||
| AK051966.1 | - | 2, 3 | 234 | |||
| AK053847.1 | - | 2, 3 | 273 | |||
| AK135394.1 | - | 2, 3 | 253 | |||
| BC050848.1 | - | 2, 3 | 214 | 6507692 | ||
| CF535627.1 | - | 2, 3 | 259 | |||

Clspn
Genomic Map