siRNA Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113360.2 | NP_001106831.1 | 22 | 4716 | ||
| NM_175556.4 | NP_780765.2 | 23 | 5247 | |||
| GenBank mRNA | AK044619.1 | - | 23 | 5249 | ||
| AK129147.1 | - | 23 | 5007 | |||
| BC036966.1 | - | 2 | 1576 | |||
| BC040465.1 | - | 10 | 4080 | |||
| BC052329.1 | - | 12 | 4202 | |||
| BC060083.1 | - | 2 | 2858 | |||
| BG261709.1 | - | 1 | 155 | |||
| CJ252783.1 | - | 1 | 143 | |||
| DQ176851.1 | - | 22 | 4716 | |||

Plch2
Genomic Map