siRNA Details
| Gene Symbol: Zfp384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252083.1 | NP_001239012.1 | 6 | 847 | ||
| NM_175557.5 | NP_780766.3 | 6 | 818 | |||
| GenBank mRNA | AK081740.1 | - | 1 | 127 | ||
| AK086185.1 | - | 6 | 743 | |||
| AK156026.1 | - | 6 | 849 | |||
| AK172476.1 | - | 4 | 1237 | |||
| BC064013.1 | - | 6 | 682 | 6515698 | ||
| DQ323640.1 | - | 1 | 77 | |||
| DQ323641.1 | - | 1 | 77 | |||

Zfp384
Genomic Map