siRNA Details
| Gene Symbol: Zfp446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168562.1 | NP_001162034.1 | 3 | 781 | ||
| NM_175558.4 | NP_780767.1 | 3 | 675 | |||
| GenBank mRNA | AK041761.1 | - | 3 | 677 | ||
| AK044666.1 | - | 2 | 1517 | |||
| AK142766.1 | - | 2 | 1170 | |||
| BC036728.1 | - | 2 | 627 | |||
| BC048909.1 | - | 3 | 617 | |||
| BC117850.1 | - | 3 | 788 | |||

Zfp446
Genomic Map