siRNA Details
| Gene Symbol: Zfp747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175560.3 | NP_780769.1 | 3 | 1599 | ||
| GenBank mRNA | AK032584.1 | - | 3 | 1565 | ||
| AK042972.1 | - | 3 | 1600 | |||
| BC058799.1 | - | 3 | 1545 | 6484930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp747| Gene Symbol: Zfp747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175560.3 | NP_780769.1 | 3 | 1599 | ||
| GenBank mRNA | AK032584.1 | - | 3 | 1565 | ||
| AK042972.1 | - | 3 | 1600 | |||
| BC058799.1 | - | 3 | 1545 | 6484930 | ||