siRNA Details
Gene Symbol: Zfp747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175560.3 | NP_780769.1 | 3 | 2526 | ||
GenBank mRNA | AK032584.1 | - | 3 | 2492 | ||
AK042972.1 | - | 3 | 2527 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175560.3 | NP_780769.1 | 3 | 2526 | ||
GenBank mRNA | AK032584.1 | - | 3 | 2492 | ||
AK042972.1 | - | 3 | 2527 |