siRNA Details
| Gene Symbol: Tmem169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175564.4 | NP_780773.1 | 3 | 1596 | ||
| GenBank mRNA | AK043681.1 | - | 3 | 1597 | ||
| AK084021.1 | - | 3 | 1577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem169| Gene Symbol: Tmem169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175564.4 | NP_780773.1 | 3 | 1596 | ||
| GenBank mRNA | AK043681.1 | - | 3 | 1597 | ||
| AK084021.1 | - | 3 | 1577 | |||