siRNA Details
| Gene Symbol: Xylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175645.3 | NP_783576.2 | 5 | 1401 | ||
| GenBank mRNA | AJ291750.1 | - | 3 | 718 | ||
| AJ539164.1 | - | 5 | 1403 | |||
| AK033309.1 | - | 5 | 1403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xylt1| Gene Symbol: Xylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175645.3 | NP_783576.2 | 5 | 1401 | ||
| GenBank mRNA | AJ291750.1 | - | 3 | 718 | ||
| AJ539164.1 | - | 5 | 1403 | |||
| AK033309.1 | - | 5 | 1403 | |||