siRNA Details
| Gene Symbol: A230050P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175687.2 | NP_783618.1 | 8 | 1571 | ||
| GenBank mRNA | AK034516.1 | - | 7 | 3020 | ||
| AK157146.1 | - | 7 | 3099 | |||
| BC031510.1 | - | 8 | 1671 | 3492596 | ||
| BC043448.1 | - | 8 | 1510 | 5151846 | ||
| BY625900.1 | - | 1 | 282 | |||

A230050P20Rik
Genomic Map