siRNA Details
Gene Symbol: A530099J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175688.4 | NP_783619.2 | 2 | 405 | ||
GenBank mRNA | AK041317.1 | - | 2 | 406 | ||
BC150908.1 | - | 2 | 284 | 9087819 | ||
BC150909.1 | - | 2 | 284 | 9087820 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
