siRNA Details
| Gene Symbol: Pianp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145926.1 | NP_001139398.1 | 6 | 1781 | ||
| NM_001145927.1 | NP_001139399.1 | 6 | 1765 | |||
| NM_175696.4 | NP_783627.2 | 6 | 1936 | |||
| GenBank mRNA | AK082984.1 | - | 6 | 1940 | ||
| AK211709.1 | - | 1 | 337 | |||
| AK216678.1 | - | 1 | 313 | |||
| AK219084.1 | - | 1 | 297 | |||
| BC065170.1 | - | 6 | 1763 | 6413408 | ||

Pianp
Genomic Map