siRNA Details
| Gene Symbol: Plxna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175750.3 | NP_786926.2 | 27, 28 | 5831 | ||
| GenBank mRNA | AB073228.1 | - | 27, 28 | 5830 | ||
| AK051614.1 | - | 12, 13 | 1734 | |||
| BC141298.1 | - | 27, 28 | 5129 | 9055910 | ||

Plxna4
Genomic Map