siRNA Details
| Gene Symbol: BLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001715.2 | NP_001706.2 | 5 | 896 | ||
| XM_011543824.1 | XP_011542126.1 | 5 | 917 | |||
| XM_011543825.2 | XP_011542127.1 | 5 | 662 | |||
| XM_011543827.1 | XP_011542129.1 | 4 | 793 | |||
| XM_011543828.2 | XP_011542130.1 | 5 | 917 | |||
| XM_011543829.2 | XP_011542131.1 | 5 | 917 | |||
| XM_017013730.1 | XP_016869219.1 | 4 | 793 | |||
| GenBank mRNA | AK313751.1 | - | 5 | 485 | ||
| BC007371.2 | - | 5 | 459 | 3634568 | ||
| BC018703.1 | - | 5 | 760 | |||
| BC032413.1 | - | 5 | 505 | 5216030 | ||
| S76617.1 | - | 5 | 903 | |||
| Z33998.1 | - | 5 | 537 | |||

BLK
Genomic Map