siRNA Details
| Gene Symbol: STOML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256672.1 | NP_001243601.1 | 7 | 1427 | ||
| NM_001256673.1 | NP_001243602.1 | 6 | 1280 | |||
| NM_001256674.1 | NP_001243603.1 | 6 | 1277 | |||
| NM_001256675.1 | NP_001243604.1 | 6 | 1217 | |||
| NM_001256676.1 | NP_001243605.1 | 7 | 1241 | |||
| NM_001256677.1 | NP_001243606.1 | 8 | 1558 | |||
| NM_001324226.1 | NP_001311155.1 | 6 | 1546 | |||
| NM_001324227.1 | NP_001311156.1 | 7 | 1244 | |||
| NM_001324228.1 | NP_001311157.1 | 7 | 1308 | |||
| NM_001324229.1 | NP_001311158.1 | 6 | 1735 | |||
| NM_001324230.1 | NP_001311159.1 | 8 | 1561 | |||
| NM_004809.4 | NP_004800.2 | 7 | 1430 | |||
| XM_005254792.4 | XP_005254849.1 | 7 | 2334 | |||
| XM_006720773.3 | XP_006720836.1 | 7 | 2332 | |||
| GenBank mRNA | AF111800.1 | - | 6 | 1229 | ||
| AF156557.1 | - | 7 | 1438 | |||
| AK075244.1 | - | 7 | 1326 | |||
| AK093689.1 | - | 3 | 1695 | |||
| AK302051.1 | - | 6 | 1235 | |||
| AL109664.1 | - | 6 | 1107 | |||
| AL109665.1 | - | 7 | 1281 | |||
| BC034379.1 | - | 7 | 1376 | 5185908 | ||
| BC065249.1 | - | 7 | 1376 | 6059393 | ||
| Y16522.1 | - | 7 | 1337 | |||

STOML1
Genomic Map