siRNA Details
| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176919.4 | NP_795893.2 | 9 | 1826 | ||
| GenBank mRNA | AK040194.1 | - | 9 | 1570 | ||
| AK040207.1 | - | 9 | 1811 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1h| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176919.4 | NP_795893.2 | 9 | 1826 | ||
| GenBank mRNA | AK040194.1 | - | 9 | 1570 | ||
| AK040207.1 | - | 9 | 1811 | |||