siRNA Details
| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176922.5 | NP_795896.4 | 3 | 314 | ||
| GenBank mRNA | AK054303.1 | - | 3 | 310 | ||
| AY124460.1 | - | 3 | 310 | |||
| BB840351.1 | - | 3 | 344 | |||
| BC058716.1 | - | 3 | 308 | 6842988 | ||

Itga11
Genomic Map