siRNA Details
| Gene Symbol: Wdr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170742.1 | NP_001164213.1 | 3 | 349 | ||
| NM_001170743.1 | NP_001164214.1 | 3 | 349 | |||
| NM_176923.4 | NP_795897.2 | 3 | 349 | |||
| GenBank mRNA | AK030644.1 | - | 3 | 230 | ||
| AK170910.1 | - | 3 | 348 | |||
| AK173301.1 | - | 3 | 312 | |||
| BC066082.1 | - | 3 | 334 | 30355918 | ||
| EF011622.1 | - | 3 | 334 | |||

Wdr59
Genomic Map