siRNA Details
| Gene Symbol: 9530053A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164655.1 | NP_001158127.1 | 5 | 1532 | ||
| GenBank mRNA | AK035473.1 | - | 5 | 1533 | ||
| AK035528.1 | - | 4 | 3640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530053A07Rik| Gene Symbol: 9530053A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164655.1 | NP_001158127.1 | 5 | 1532 | ||
| GenBank mRNA | AK035473.1 | - | 5 | 1533 | ||
| AK035528.1 | - | 4 | 3640 | |||