siRNA Details
| Gene Symbol: Dennd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162917.1 | NP_001156389.1 | 5 | 860 | ||
| GenBank mRNA | AK089365.1 | - | 6 | 827 | ||
| AK158187.1 | - | 5 | 700 | |||
| AK158279.1 | - | 5 | 759 | |||
| BC172087.1 | - | 4 | 518 | 9054373 | ||

Dennd4a
Genomic Map