siRNA Details
| Gene Symbol: Nwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176940.5 | NP_795914.3 | 7 | 2295 | ||
| GenBank mRNA | AK034868.1 | - | 1994 | |||
| AK038792.1 | - | 6 | 1988 | |||
| AK087622.1 | - | 7 | 2300 | |||
| AK162888.1 | - | 7 | 2290 | |||
| BC082552.1 | - | 7 | 2235 | |||
| BC144786.1 | - | 5 | 1885 | 9053334 | ||
| BC144787.1 | - | 5 | 1885 | |||
| BC145703.1 | - | 5 | 1885 | 40131006 | ||

Nwd1
Genomic Map