siRNA Details
| Gene Symbol: 9630028B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036028.1 | - | 2 | 715 | ||
| AK048735.1 | - | 2 | 793 | |||
| BC147657.1 | - | 2 | 693 | 9087446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9630028B13Rik| Gene Symbol: 9630028B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036028.1 | - | 2 | 715 | ||
| AK048735.1 | - | 2 | 793 | |||
| BC147657.1 | - | 2 | 693 | 9087446 | ||